TaqMan™ Genotyping Master Mix
Actual product may vary
TaqMan™ Genotyping Master Mix
TaqMan™ Genotyping Master Mix
Applied Biosystems™

TaqMan™ Genotyping Master Mix

**********************Alternative Product: Try TaqPath ProAmp Master Mix, our newest and highest performing master mix for genotyping and copy number variationRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
43816562 x 10 mL
43713531 x 1 mL
43713551 x 10 mL
43713571 x 50 mL
43816572 x 50 mL
Catalog number 4381656
Price (USD)
2,056.65
Online exclusive
2,285.00
Save 228.35
Each
Quantity:
2 x 10 mL
Price (USD)
2,056.65
Online exclusive
2,285.00
Save 228.35
Each
**********************
Alternative Product: Try TaqPath ProAmp Master Mix, our newest and highest performing master mix for genotyping and copy number variation for accurate results, even from samples containing PCR inhibitors. With TaqPath ProAmp Master Mix, we’ve taken the best of TaqMan Genotyping Master Mix and added additional capabilities for your genotyping and copy number variation experiments.

**********************
TaqMan™ Genotyping Master Mix is optimized for end-point fluorescence detection in SNP genotyping applications.

• Distinct clusters and high call rates for unambiguous allelic discrimination
• Validated with TaqMan™ SNP Genotyping Assays
• Excellent pre- and post-PCR stability for high throughput setup and analysis

May be used for a variety of genotyping applications, including:

• Candidate gene studies
• Drug-target validation
• Disease association studies
• Population genetics
• Linkage mapping
• Agricultural applications

Reaction size is based on a 50 μl reaction volume; please see below for other reaction sizes.

Exceptional Performance from an Optimized Mix (2X)
Accurate genotype assignments result from preferential binding of the allele-specific probe to the matching target. TaqMan™ Genotyping Master Mix enables specific binding of the probe to achieve exceptional cluster resolution. Key features include:

• AmpliTaq Gold™ DNA Polymerase, UP (Ultra Pure) – Automatic hot start enzyme designed to be active during thermal cycling and inactive at room temperature for easy reaction setup.
• Optimized mix components provide excellent specificity for discrimination between alleles
• Passive internal reference based on proprietary ROX™ dye for increased precision on Applied Biosystems™ real-time PCR instruments
• Single thermal cycling condition for consistent results with TaqMan™ Assays

Validated with TaqMan™ Genotyping Assays and Instruments
TaqMan™ Genotyping Master Mix has been tested across all types of TaqMan™ SNP Genotyping Assays: TaqMan™ Drug Metabolism Genotyping Assays, TaqMan™ SNP Genotyping Assays, and Custom TaqMan™ SNP Genotyping Assays. In addition, the master mix is validated with the Applied Biosystems™ thermal cyclers and real-time PCR systems.

High-Throughput Setup and Analysis
The combination of several low-cost thermal cyclers for PCR and a single real-time PCR instrument for allelic discrimination make high throughput SNP genotyping manageable. TaqMan™ Genotyping Master Mix further improves high-throughput setup and analysis with excellent room temperature stability both before and after PCR. This provides the flexibility required to run long experiments, unattended overnight or over a weekend.

Reliable Discrimination with Challenging Targets
Amplicon design constraints make SNP detection challenging, particularly when targets contain an abundance of GC- or AT-rich regions. The optimized components of TaqMan™ Genotyping Master Mix result in tight, well-separated clusters and more accurate allele-calls compared to other mixes, even for challenging targets.

Setting the Industry Standard for Quality
Extensive analytical and functional tests are performed for each manufactured lot of TaqMan™ Genotyping Master Mix, including one TaqMan™ SNP Genotyping Assay and one TaqMan™ Drug Metabolism Genotyping Assay. Results are compiled in an informative Certificate of Analysis, which is accessible through the Applied Biosystems™ Web site (search for 'Certificate of Analysis').

Number of Genotyping Reactions at a Given Reaction Size:

4371355, TaqMan™ Genotyping Master Mix, 1-Pack (1 x 10 mL)
• 50 μL Reactions: 400
• 20 μL Reactions: 1000
• 10 μL Reactions: 2,000
• 5 μL Reactions: 4,000

4381656, TaqMan™ Genotyping Master Mix, 2-Pack (2 x 10 mL)
• 50 μL Reactions: 800
• 20 μL Reactions: 2,000
• 10 μL Reactions: 4,000
• 5 μL Reactions: 8,000

4371357, TaqMan™ Genotyping Master Mix, (1 x 50 mL)
• 50 μL Reactions: 2000
• 20 μL Reactions: 5,000
• 10 μL Reactions: 10,000
• 5 μL Reactions: 20,000

4381657, TaqMan™ Genotyping Master Mix, (2 x 50 mL)
• 50 μL Reactions: 4000
• 20 μL Reactions: 10,000
• 10 μL Reactions: 20,000
• 5 μL Reactions: 40,000

Note: See user's manual or package insert for limited label license, and trademark information. For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)2 X
For Use With (Equipment)Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7
Genotyping TargetCopy Number Variants, SNPs (Unknown or Numerous), Drug Metabolizing Enzymes, SNPs (Known)
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq Gold DNA Polymerase
Product LineTaqMan
Product TypeGenotyping Master Mix
Purity or Quality GradeUP (Ultra Pure)
Quantity2 x 10 mL
Sample TypeDNA (Genomic)
TechniqueSNP Genotyping (Real-Time PCR-Based), CNV (Copy Number Variation)
Volume20 mL
Concentration2X
Detection MethodPrimer-probe
For Use With (Application)Genotyping
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Supplied at 2X concentration. The mix is optimized for SNP genotyping using TaqMan™ probes and contains AmpliTaq Gold™ DNA Polymerase UP (Ultra Pure), dNTPs without dUTP, Passive Reference 1 and optimized mix components. This pack contains two 10 mL bottles, sufficient for 800 reactions at the 50 μL total volume.

Storage: 2-8°C

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Frequently asked questions (FAQs)

What master mix can I use with the TaqMan SNP Genotyping Assays?

We recommend using either the TaqPath ProAmp Master Mix or the TaqMan Genotyping Master Mix. The TaqMan Genotyping Master Mix has the advantage of proven performance with up to 3 days of pre- and post-PCR stability, allowing you to set up plates ahead of time or read the plates later (see the data here, https://tools.thermofisher.com/content/sfs/brochures/cms_039236.pdf) while the TaqPath ProAmp Master Mix can handle samples that may have inhibitors present.

What can I do to improve the sensitivity of my qPCR assay?

If you are targeting a low-abundance gene, you may have trouble getting Ct values in a good, reliable range (Ct > 32). To increase the sensitivity of the assay, you may want to consider the following:

- Increase the amount of RNA input into your reverse transcription reaction, if possible
- Increase the amount of cDNA in your qPCR reaction (20% by volume max)
- Try a different reverse transcription kit, such as our SuperScript VILO Master Mix, for the highest cDNA yield possible
- Consider trying a one-step or Cells-to-CT type workflow (depending on your sample type)

How do I set the baseline for my qPCR experiment?

Most times your instrument software can automatically set a proper baseline for your data. Check out our short video, Understanding Baselines, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=5BjFAJHW-bE).

How do I set the threshold for my qPCR experiment?

In most cases your instrument software can automatically set a proper threshold for your data. Check out our short video, Understanding Thresholds, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=H_xsuRQIM9M).

I am not getting any amplification with my TaqMan Assay or SYBR Green primer set. What is causing this?

There could be several reasons for no amplification from an assay or primer set. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/no-amplification.html) for more details.