Methyl 4-bromo-3-methylbenzoate, 95%
Methyl 4-bromo-3-methylbenzoate, 95%
Methyl 4-bromo-3-methylbenzoate, 95%
Thermo Scientific Chemicals

Methyl 4-bromo-3-methylbenzoate, 95%

CAS: 148547-19-7 | C9H9BrO2 | 229.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 438190250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS148547-19-7
IUPAC Namemethyl 4-bromo-3-methylbenzoate
Molecular FormulaC9H9BrO2
InChI KeyGTZTYNPAPQKIIR-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(Br)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to yellow
Melting point38°C to 44°C
Appearance (Form)Low melting solid or liquid
GC>=94.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only