Product Image
Applied Biosystems™

TaqMan™ Francisella tularensis Detection Kit

Applied Biosystems™ TaqMan™ Francisella tularensis Detection Kit uses the polymerase chain reaction (PCR) to amplify a unique microorganism specific DNARead more
Have Questions?
Catalog NumberQuantity
4382490100 reactions
Catalog number 4382490
Price (USD)
2,195.00
Each
Quantity:
100 reactions
Price (USD)
2,195.00
Each
Applied Biosystems™ TaqMan™ Francisella tularensis Detection Kit uses the polymerase chain reaction (PCR) to amplify a unique microorganism specific DNA target sequence and a TaqMan™ probe to detect the amplified sequence. The TaqMan™ Francisella tularensis Detection Kit provides a simple, reliable, and rapid procedure that utilizes the polymerase chain reaction (PCR) to amplify a target sequence unique to Francisella tularensis
• Reduce sample handling with a 3-step process that uses a single, closed-tube format during the PCR step.
• Highly sensitive TaqMan™ Real-Time PCR system enables a dramatic reduction in time required for pre-enrichment compared to conventional techniques.
• Streamline your workflow with one standard protocol for multiple organisms.
3 Easy Steps
This kit has 3 easy steps: enrich samples, prepare samples, and run samples. Grow the specific pathogen from a selected sample using a standard enrichment procedure in the enrichment step. In the sample preparation step isolate the DNA for PCR, and prepare, mix and pipet the reactions. In the final step, analyze your sample by running a plate on one of our real-time PCR systems.

A Complete Solution
We offer a full line of TaqMan™ Pathogen Detection kits based on universal assay conditions, enabling you to run any combination of these assays together in the same sample run. In combination with our real-time PCR systems, the TaqMan™ Francisella tularensis Detection Kit offers a fully integrated, single-vendor solution to identify Francisella tularensis
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DiseaseTularemia
FormatTube
No. of Reactions100 Reactions
Product LineTaqMan
Product TypeFrancisella Tularensis Detection Kit
Quantity100 reactions
Shipping ConditionDry Ice
TargetBacteria
Target Organism ClassF. tularensis [Tularemia]
Target SpecificityTarget-Specific
Detection MethodPrimer-probe
For Use With (Application)Pathogen Detection
GC-Rich PCR PerformanceHigh
Label or DyeFAM
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
300μL 10X Francisella tularensis Target Assay Tul4 (store at -20°C), 300μL 10X Francisella tularensis Target Assay fopA (store at -20°C), 1,000μL Negative Control (store at 2°C-8°C), Two Tubes 2X Fast PCR Master Mix 1,500μL (store at 2°C-8°C).