BioProbes® Journal, ProbesOnline™, and BioPath™ E-newsletters. Subscribe today!

Streptavidin-FITC (ZyMax Grade)

Catalog number: 438311

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Reactivity: FITC-SA reacts with biotin.

Applications: Immunofluorescence, Western blotting, and ELISA

ZyMAX™ reagents offer enhanced assay sensitivity, higher titer, and greater flexibility in assay design than standard grade reagents.

Documents

Manuals & protocols