Isobutanesulfonyl chloride, 97%
Isobutanesulfonyl chloride, 97%
Isobutanesulfonyl chloride, 97%
Thermo Scientific Chemicals

Isobutanesulfonyl chloride, 97%

CAS: 35432-36-1 | C4H9ClO2S | 156.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 438410010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS35432-36-1
IUPAC Name2-methylpropane-1-sulfonyl chloride
Molecular FormulaC4H9ClO2S
InChI KeyNQLNHTOCFWUYQE-UHFFFAOYSA-N
SMILESCC(C)CS(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear colorless to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only