Invitrogen™

Human Heart Mitochondria WB Control, unconjugated

Catalog number:  438505

 Related applications: Cell Structure | Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Western blot control for human heart mitochondria.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Size: 50 µg
Species: Human
Shipping Condition: Wet Ice

Contents & storage

Shipped at 4°C. Store at -20°C after receipt or -80°C for long term.

Documents

Manuals & protocols

Recommended products