Product Image
Applied Biosystems™
Variant Reporter™ Software v1.1, 30 Day Demo
The Variant Reporter™ Software performs comparative sequencing, also known as direct sequencing, medical sequencing, PCR sequencing and resequencing with DNA sequencing files.
Have Questions?
카탈로그 번호수량
43852701 CD
카탈로그 번호 4385270
제품 가격(KRW)
126,000
1 CD
수량:
1 CD
제품 가격(KRW)
126,000
1 CD
The Variant Reporter™ Software performs comparative sequencing, also known as direct sequencing, medical sequencing, PCR sequencing and resequencing with DNA sequencing files. The software is designed for reference based and non-reference based analysis such as mutation detection and analysis, SNP discovery and validation and sequence confirmation. The robust algorithms will call SNPs, mutations, insertions, deletion and heterozygous insertions⁄deletions for data generated using the Applied Biosystems™ Genetic Analyzers.
There are two ways to obtain the 30-day demo software
  1. Register and download the free demo Variant Reporter™ Software: http://marketing.appliedbiosystems.com/mk/get/VARIANT_REPORTER0507_LOGIN?isource=fr_E_AB_Product_Page
  2. Add the 30-day demo CD (part number 4385270) to your shopping basket and it will be shipped to you for free.
For Research Use Only. Not for use in diagnostic procedures.
사양
데이터 입력Sequencing traces
데이터 출력Variant Reports with sequence comparison metrics
용도(장비)310 Genetic Analyzer, 3100 Genetic Analyzer, 3100-Avant Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer, 3500 Genetic Analyzer, 3730 DNA Analyzer
라이선스Demo
운영 체제Windows Vista Business, Windows XP Pro
제품 유형Variant Reporter Software V1.1
수량1 CD
소프트웨어 카테고리Software
시스템 요구 사항Computer: Core 2 Duo E8400/3 GHz; 2 GB RAM
제품라인Variant Reporter
Software TypeComparative Sequencing
Unit Size1 CD
구성 및 보관
Contains one software CD.
자주 묻는 질문(FAQ)
We are upgrading to the Windows 10 operating system. What are the compatible capillary electrophoresis secondary analysis software with Windows 10?
The following capillary electrophoresis secondary analysis software are compatible with Windows 10 (and also Windows 7):
-Sequencing Analysis Software v7
-SeqScape Software v4
-Variant Reporter Software v3
-GeneMapper Software v6
-Minor Variant Finder v1.2
-MicrobeBridge v1.1
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Where do I get the latest Applied Biosystems Capillary Electrophoresis Software patches?
Please visit our Applied Biosystems Genetic Analyzer Freeware and Patches website: https://www.thermofisher.com/sangerpatches
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How can I import my data from Variant Reporter Software?
For importing data, there are many ways to do so:
More information on backing up the Data Store can be found in the Variant Reporter Help Contents under the keyword “Back-up”.
1. Select the Data Store where you would like the data imported to. To choose a Data Store:
a. Click the Manage button in the Data Store section of the Dashboard View.
b. Highlight the Data Store you wish to use and click on Change Location to select it. Click Close to close the window.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How can I archive a selected project in the Variant Reporter Software?
To archive a Variant Reporter software project from the Dashboard:
1. Open the Dashboard View.
2. In the Project tab select the project you want to archive, then click .
3. In the Archive Destination dialog box, browse to the folder where you want the project archived.
4. Click OK. The project file you archived will no longer appear in the Variant Reporter Dashboard.
More information on archiving Project data can be found in the Variant Reporter Help Contents under the keyword “Archive”.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How can I back up the Data Store in the Variant Reporter Software?
To back up the Data Store:
1. Open the Dashboard View.
2. Click .
3. In the Browse for Back-up Folder Destination dialog box, select the folder that you want to back up the Data Store to, then click OK. Your project will start to get backed up.
Note: All associated Projects, Analysis Parameters, and References contained in the current Data Store will be copied to the folder you selected when you back up the Variant Reporter Data Store.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.