Path-ID™ qPCR Master Mix
Applied Biosystems™

Path-ID™ qPCR Master Mix

Ambion™ Path-ID™ qPCR Master Mix is a convenient enzyme, buffer, and dNTP mix for TaqMan™ probe-based, real-time PCR detection ofRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
4388643100 Reactions
4388644500 Reactions
Catalog number 4388643
Price (USD)
221.65
Online exclusive
246.00
Save 24.35
Each
No. of Reactions:
100 Reactions
Price (USD)
221.65
Online exclusive
246.00
Save 24.35
Each
Ambion™ Path-ID™ qPCR Master Mix is a convenient enzyme, buffer, and dNTP mix for TaqMan™ probe-based, real-time PCR detection of DNA targets. It is optimized for targets of importance in animal health and to function in the presence of PCR inhibitors. The mix is supplied in one tube suitable for 100 reactions. With the exception of PCR primers and probes for your target, the master mix contains everything you need for real-time PCR, including:
• AmpliTaq Gold™ ultrapure thermostable DNA polymerase designed for automatic hot start, to minimize nonspecific PCR products
• Optimized buffer and dNTP concentration, for reliable PCR
• Passive internal reference, based on proprietary ROX™ dye, for precise data analysis.
For Veterinary Use Only.
Specifications
For Use With (Equipment)7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode
FormatTube
Hot StartBuilt-In Hot Start
No. of Reactions100 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq Gold DNA Polymerase
Product LineAmbion, Path-ID
Product TypeqPCR Master Mix
Purity or Quality GradeUP (Ultra Pure)
Quantity100 reactions
Sample TypeDNA (Genomic), cDNA
Shipping ConditionDry Ice
Sufficient For100 Reactions
Volume1.375 mL
Concentration2X
Detection MethodPrimer-probe
For Use With (Application)Animal Health, Pathogen Detection
FormLiquid
PCR MethodqPCR
Reaction SpeedFast or Standard
Unit SizeEach
Contents & Storage
2X qPCR Master Mix is stored at –20°C. Nuclease-free water may be stored at any temperature.