2,3-Diphenylmaleic anhydride, 98%
2,3-Diphenylmaleic anhydride, 98%
2,3-Diphenylmaleic anhydride, 98%
Thermo Scientific Chemicals

2,3-Diphenylmaleic anhydride, 98%

CAS: 4808-48-4 | C16H10O3 | 250.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 439010250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS4808-48-4
IUPAC Namediphenyl-2,5-dihydrofuran-2,5-dione
Molecular FormulaC16H10O3
InChI KeyOUJCFCNZIUTYBH-UHFFFAOYSA-N
SMILESO=C1OC(=O)C(=C1C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Yellow
Melting point156°C to 162°C
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only