Cyclopropanesulfonyl chloride, 97%
Cyclopropanesulfonyl chloride, 97%
Cyclopropanesulfonyl chloride, 97%
Thermo Scientific Chemicals

Cyclopropanesulfonyl chloride, 97%

CAS: 139631-62-2 | C3H5ClO2S | 140.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 439020010
Price (TWD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS139631-62-2
IUPAC Namecyclopropanesulfonyl chloride
Molecular FormulaC3H5ClO2S
InChI KeyPFWWSGFPICCWGU-UHFFFAOYSA-N
SMILESClS(=O)(=O)C1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only