SurePac™ Bio 550 SEC MDi™ HPLC Columns
SurePac™ Bio 550 SEC MDi™ HPLC Columns
SurePac™ Bio 550 SEC MDi™ HPLC Columns
Thermo Scientific™

SurePac™ Bio 550 SEC MDi™ HPLC Columns

Experience the future of Size-Exclusion Chromatography (SEC) with SurePac™ Bio 550 SEC MDi™ analytical columns and guards exclusively designed to deliver robust and reproducible performance at high resolution and throughput to accelerate your therapeutics analysis process.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoLength (Metric)Diameter (Metric)Column FormatFlow Rate
43903-03783130 mm7.8 mmGuard Column1.0 mL/min.
43903-152131150 mm2.1 mmAnalytical Column125 μL/min.
43903-154631150 mm4.6 mmAnalytical Column0.6 mL/min.
43903-157831150 mm7.8 mmAnalytical Column1.0 mL/min.
43903-03213130 mm2.1 mmGuard Column125 μL/min.
43903-03463130 mm4.6 mmGuard Column0.6 mL/min.
Número do catálogo 43903-037831
Preço (BRL)
-
Length (Metric):
30 mm
Diameter (Metric):
7.8 mm
Column Format:
Guard Column
Flow Rate:
1.0 mL/min.

Experience the future of Size-Exclusion Chromatography (SEC) with SurePac™ Bio 550 SEC MDi™ analytical columns and guards exclusively designed to deliver robust and reproducible performance at high resolution and throughput to accelerate your therapeutics analysis process. The column's groundbreaking MDi™ technology integrates our advanced 3 μm monodisperse silica particle platform with bioinert hardware to ensure superior reliability for best sample recovery. SurePac™ Bio 550 SEC MDi™ column is ideal for AAV gene therapeutics analysis and offers excellent selectivity due to the dedicated smaller particle (3 μm) size combined with wide-pore (550Å) for separating monomeric AAVs from aggregates based on their size differences.

SurePac Bio 550 SEC MDi columns and guards are based on high-purity, spherical, porous (550 Å), 3 μm monodispersed silica particles that are covalently modified with a proprietary diol hydrophilic layer. This unique design yields shorter diffusion distances and enhanced reproducible mass transfer and narrower peaks. The consistent size of these particles facilitates precise control over media synthesis and column packing, significantly improving column-to-column and lot-to-lot reproducibility. The columns are housed in a hydrophilic bioinert-coated stainless-steel hardware to eliminate secondary interactions, ensuring optimal performance during the initial injection process.

  • Ideal for AAV gene therapy characterization and quality control assessment due to selectivity of the dedicated smaller particle (3 μm) combined with wide-pore (550Å) size
  • Superior column-to-column and lot-to-lot reproducibility eliminates column variability as a concern in stability and QA/QC testing
  • High-efficiency peaks with excellent recovery
  • Higher resolution and throughput for fast turnaround time without sacrificing performance
  • Bioinert hardware to eliminate analyte adsorption
Especificações
Column FormatGuard Column
Flow Rate1.0 mL/min.
Packing MaterialMonodisperse Silica Resin
Quantity1 Ea.
Stationary PhaseProprietary
Temperature20°C to 60°C
Diameter (Metric)7.8 mm
Length (Metric)30 mm
Particle Size3 μm
PhaseSize Exclusion
TechniqueSEC
pH2 to 8
Unit SizeEach