N-epsilon-FMOC-N-alpha-BOC-L-Lysine, 99%
N-epsilon-FMOC-N-alpha-BOC-L-Lysine, 99%
N-epsilon-FMOC-N-alpha-BOC-L-Lysine, 99%
Thermo Scientific Chemicals

N-epsilon-FMOC-N-alpha-BOC-L-Lysine, 99%

CAS: 84624-27-1 | C26H32N2O6 | 468.55 g/mol
Have Questions?
Catalog NumberQuantity
4390800505 g
Catalog number 439080050
Price (EUR)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS84624-27-1
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-6-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Molecular FormulaC26H32N2O6
InChI KeyJYEVQYFWINBXJU-ANBDAQEENA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation+2.3° to +4.3° (20°C, 589 nm) (c=1 in Ethyl acetate)
Infrared spectrumConforms
Appearance (Color)White
Appearance (Form)Powder
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only