Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0), 96%
Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0), 96%
Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0), 96%
Thermo Scientific Chemicals

Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0), 96%

CAS: 811862-77-8 | C42H44O10Pd | 815.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 439150010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS811862-77-8
IUPAC Namebis(1,5-bis(3,5-dimethoxyphenyl)penta-1,4-dien-3-one) palladium
Molecular FormulaC42H44O10Pd
InChI KeyRTGAJOPJZJDWAX-UHFFFAOYSA-N
SMILES[Pd].COC1=CC(C=CC(=O)C=CC2=CC(OC)=CC(OC)=C2)=CC(OC)=C1.COC1=CC(C=CC(=O)C=CC2=CC(OC)=CC(OC)=C2)=CC(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red-brown
Infrared spectrumConforms
NMR>=95.0 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only