PrepFiler® Spin Tubes and Filter Columns (300 tubes/100 columns)
Catalog number: 4392342
Applied Biosystems™  Related applications: DNA Extraction | Forensics
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The PrepFiler® Spin Tubes and Filter Columns are now available for purchase separately to provide the ordering flexibility to select only the plastics consumables needed for use with all PrepFiler® Forensic DNA Extraction Kits. The full PrepFiler® Forensic DNA Extraction Kits use magnetic particles with an optimized multi-component surface chemistry to deliver superior DNA yield from most forensic sample types, including body fluids, and stains and swabs of body fluids.
The PrepFiler® Filter Columns are used for substrate removal and the PrepFiler® Spin Tubes are used for sample processing in tube format.
The full PrepFiler® Forensic DNA Extraction Kit is also available for purchase, and contains other components required for DNA recovery from both routine and challenging samples, including removal of PCR inhibitors.
For Forensic or Paternity Use Only.
Specifications
Product Line: Applied Biosystems™, PrepFiler®
Product Size: 100 columns
Shipping Condition: Room Temperature
Contents & storage
Store tubes and columns at ambient temperature.
Documents
Manuals & protocols