SOLiD™ Deposition Chambers, 8 wells

Catalog number: 4392751

Applied Biosystems™  Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This is an accessory for use with the SOLiD™ Analyzer.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): SOLiD™ 3 Analyzer, SOLiD™ 4 System
Format: 8-wells
Product Line: SOLiD™
Product Size: 2 each
Shipping Condition: Room Temperature
Type: Deposition Chambers

Contents & storage

Store at room temperature. Two additional deposition chambers for 8 samples⁄slide.

Documents

Manuals & protocols