What is the white residue around the Applied Biosystems 3500/3500xL Genetic Analyzer array knob?
When placing the array on the instrument, the last step in the process is to loosen the array knob and the instrument forces any air bubbles around the array tip through the threading and the middle of the array knob. Some polymer gets pushed through as well and when it dries up, it appears as a white, flaky residue. Use a damp, lint-free cloth to remove it.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.