2-Chloro-1,3-dimethylimidazolinium chloride, 90%
2-Chloro-1,3-dimethylimidazolinium chloride, 90%
2-Chloro-1,3-dimethylimidazolinium chloride, 90%
Thermo Scientific Chemicals

2-Chloro-1,3-dimethylimidazolinium chloride, 90%

CAS: 37091-73-9 | C5H10Cl2N2 | 169.05 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
產品號碼 439520250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS37091-73-9
IUPAC Name2-chloro-1,3-dimethyl-4,5-dihydro-1H-imidazol-3-ium chloride
Molecular FormulaC5H10Cl2N2
InChI KeyAEBBXVHGVADBHA-UHFFFAOYSA-M
SMILES[Cl-].CN1CC[N+](C)=C1Cl
檢視更多
規格Specification Sheet規格表
Infrared spectrumConforms
Elemental Analysis31.26 to 39.79 % (C)
Appearance (Form)Crystalline powder or solid
HPLC>=88.0 %
Elemental Analysis14.58 to 18.56 % (N)
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only