N-Isopropylmethylamine, 98%
N-Isopropylmethylamine, 98%
N-Isopropylmethylamine, 98%
Thermo Scientific Chemicals

N-Isopropylmethylamine, 98%

CAS: 4747-21-1 | C4H11N | 73.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Catalog number 439650500
Price (USD)
231.65
Special offer
Online exclusive
Ends: 28-Dec-2026
272.00
Save 40.35
Each
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
231.65
Special offer
Online exclusive
Ends: 28-Dec-2026
272.00
Save 40.35
Each
Chemical Identifiers
CAS4747-21-1
IUPAC Namemethyl(propan-2-yl)amine
Molecular FormulaC4H11N
InChI KeyXHFGWHUWQXTGAT-UHFFFAOYSA-N
SMILESCNC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.3830 to 1.3850 (20°C, 589 nm)
Titration with HClO4>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only