Do you need this panel in a 384-well plate? Please visit Specialty Plating to learn more.
Applied Biosystems™
TaqMan™ Array Human Endogenous Control
Applied Biosystems™ TaqMan™ Express Endogenous Control Plates use TaqMan™ probe-based chemistry and are designed for use on the suite ofRead more
Have Questions?
Catalog Number
Quantity
4396840
1 standard plate
Catalog number 4396840
Price (USD)
271.00
Each
Quantity:
1 standard plate
Price (USD)
271.00
Each
Applied Biosystems™ TaqMan™ Express Endogenous Control Plates use TaqMan™ probe-based chemistry and are designed for use on the suite of Applied Biosystems™ Real-Time PCR Systems – together the gold standard in quantitative gene expression offering the greatest sensitivity, specificity, reproducibility, and the broadest dynamic range. The first aspect of any experiment looking at relative quantitation of gene expression should be the selection of endogenous control gene(s), to normalize for variations in sample input. The TaqMan™ Express Human Endogenous Control Plate contains 32 genes, plated in triplicate, that have been shown to be good candidates for such an experiment. This collection of genes has been selected from literature searches and⁄or whole genome microarray tests carried out on numerous human tissues. They have been shown to be expressed constitutively and at moderate abundance across most test samples.
For Research Use Only. Not for use in diagnostic procedures.
What is the minimum amount of TaqMan array plates I need to purchase to submit an order?
The minimum amount of TaqMan Array plates needed to submit an order is based on the plate type:
• Inventoried TaqMan array plates: 1 plate
• TaqMan Flexible Content panel array plates: 6 plates
• Custom TaqMan array plates: 6 plates
• Specialty TaqMan array plates: 10-20 plates (depends on assay format and configuration). Please contact our Specialty Plates team at Specialty_Plates@thermofisher.com for details.
Can our TaqMan Array Plates be run on non-Applied Biosystem real-time PCR instruments?
We recommend using our Plastics Selection Tool to verify which plates are compatible with your instrument model, https://www.thermofisher.com/us/en/home/life-science/pcr/pcr-plastics/plastics-selection-guide.html.
Based on our testing, the 96-well TaqMan Array Plates can be used on the following non-Applied Biosystem real-time PCR instruments:
• Standard (0.2mL) 96-well Plates: Bio-Rad IQ5
• Fast (0.1mL) Plates: Bio-Rad CFX Connect and Bio-Rad CFX96 Touch
How do I obtain TaqMan documentation files for my TaqMan array plates?
Beginning in April 2016, TaqMan documentation files for TaqMan Array Plates (assay information files (AIF), experimental setup files, and plate layout files) can be downloaded from the Thermo Fisher Scientific website at https://www.thermofisher.com/taqmanfiles. To obtain your TaqMan documentation files:
• Select the appropriate product type.
• Enter the information being requested for the product type.
• Fixed TaqMan array plates: Enter the Part number and Batch number
• TaqMan Flexible Content panel array plates/Custom TaqMan array plates: Enter the Sales Order number and Batch number
• Click Submit.
Can poly A+ purified RNA samples be used with the TaqMan Human Endogenous Control Plate (Cta. No. 4309199)?
No. The plate has been designed to evaluate only total RNA. Ribosomal RNA will be removed from poly A+ purified RNA samples, and is required for accurate detection of the 18s rRNA endogenous control assay used on the plate.
What types of templates can be used with the TaqMan Human Endogenous control Plate (P/N 4396840)?
The plate amplifies mRNA contained within total RNA. Non-human templates are not recommended, as all the assays are human specific with the exception of 18S rRNA and the internal positive control. Additionally, Poly A+ RNA cannot be accurately evaluated by the 18S rRNA assay as most of the rRNA has been removed.