Lithium acetylide ethylenediamine complex, 25 wt.% slurry in toluene, AcroSeal™
Lithium acetylide ethylenediamine complex, 25 wt.% slurry in toluene, AcroSeal™
Lithium acetylide ethylenediamine complex, 25 wt.% slurry in toluene, AcroSeal™
Lithium acetylide ethylenediamine complex, 25 wt.% slurry in toluene, AcroSeal™
Thermo Scientific Chemicals

Lithium acetylide ethylenediamine complex, 25 wt.% slurry in toluene, AcroSeal™

CAS: 6867-30-7 | C4H9LiN2 | 92.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
439881000100 mLAcroSeal Glass Bottle
Catalog number 439881000
Price (USD)
-
Quantity:
100 mL
Packaging:
AcroSeal Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS6867-30-7
IUPAC Nameethane-1,2-diamine; ethynyllithium
Molecular FormulaC4H9LiN2
InChI KeyQJQWXTYPTBEPGS-UHFFFAOYSA-N
SMILES[Li]C#C.NCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Tan to orange-brown
Appearance (Form)Slurry
Titration with HClO423.5 to 30.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only