AmpliTaq Gold™ 360 DNA Polymerase
Applied Biosystems™

AmpliTaq Gold™ 360 DNA Polymerase

The Applied Biosystems AmpliTaq Gold 360 DNA Polymerase, when used with improved AmpliTaq Gold 360 Buffer, and the optional 360 GC Enhancer, amplifies a vast range of DNA sequence contexts.
Have Questions?
變更視圖buttonViewtableView
產品號碼Quantity
4398823250 Units
4398813100 Units
43988331000 Units
439889825 x 1000 Units
43988921500 Units
43988965000 Units
產品號碼 4398823
價格 (TWD)
9,450.00
線上優惠
Ends: 30-Sep-2026
12,600.00
您節省 3,150.00 (25%)
Each
Quantity:
250 Units
Request bulk or custom format
價格 (TWD)
9,450.00
線上優惠
Ends: 30-Sep-2026
12,600.00
您節省 3,150.00 (25%)
Each
The Applied Biosystems AmpliTaq Gold 360 DNA Polymerase, when used with AmpliTaq Gold 360 Buffer and the optional 360 GC Enhancer, amplifies a vast range of DNA sequence contexts. This polymerase efficiently amplifies targets present at low copy number, even in the presence of high concentrations of complex DNA, making it especially suited for low-copy pathogen detection and amplification of targets from degraded DNA samples. AmpliTaq Gold 360 DNA Polymerase delivers 360° coverage for a full range of targets.

AmpliTaq Gold 360 DNA Polymerase is purified by a proprietary separation process to eliminate contaminating bacterial DNA sequences from the enzyme preparation. When used with the enhanced 360 Gold Buffer, this ultra-pure enzyme, in addition to its hot-start capabilities, reduces false positive results, amplifies low-level target sequences, and promotes the amplification of a variety of templates including those from bacterial and human genomes.

Features

  • Optimized for the broadest range of targets
  • High sensitivity, specificity, and yield
  • Robust amplification of GC-rich sequences with 360 GC Enhancer
  • High-quality sequencing data
  • Efficiently and reproducibly amplifies sequences up to 5 Kb

Optimized for easy and challenging targets

Challenging targets include AT-rich, GC-rich, primer-dimer forming amplicons, homopolymer repeats, and amplicons that pose sequencing challenges. Amplicons that previously required specialized enzymes and reaction conditions can now be reproducibly amplified with a single reagent under standardized conditions (see figure).

Optimized for hot-start PCR

AmpliTaq Gold 360 DNA Polymerase provides the same hot-start specificity as AmpliTaq Gold DNA Polymerase. A high-temperature incubation step is required to activate AmpliTaq Gold DNA Polymerase, which ensures that the active enzyme is generated only at temperatures in which the DNA is fully denatured and when the primers are not annealed.

When the polymerase is added to the reaction mixture at room temperature, primer extension does not occur because the enzyme is inactivated. Any low stringency mispriming events that may have occurred will not be enzymatically extended and will not be amplified. Hence, PCR setup can be performed at room temperature without concern for extension at misprimed sites. The amount of AmpliTaq Gold 360 DNA Polymerase increases in the reaction slowly with each cycle number, and specific product yield increases without buildup of nonspecific products, including primer dimers.

Notes

  • See figure below for a demonstration of robust PCR amplification of long human and plasmid DNA.
  • See user's manual or package insert for limited label license and trademark information.
For Research Use Only. Not for use in diagnostic procedures.
規格
Exonuclease Activity5' - 3'
Fidelity (vs. Taq)1X
FormatTube
Hot StartBuilt-In Hot Start
No. of Reactions200 Reactions
Overhang3'-A
PolymeraseAmpliTaq Gold 360 DNA Polymerase
Product TypeDNA Polymerase
Quantity250 Units
Reaction FormatSeparate Components
Shipping ConditionRoom Temp or Wet Ice
Size (Final Product)5 kb or less
Starting MaterialDNA
Concentration5 U/μL
For Use With (Application)Hot-start PCR
GC-Rich PCR PerformanceHigh
Reaction SpeedStandard
Unit SizeEach
內容物與存放
• AmpliTaq Gold 360 DNA Polymerase (5 U/μL), 1 x 50 μL
• 10X AmpliTaq Gold 360 Buffer, 1 x 1.5 mL
• 25 mM MgCl2, 1 x 1.5 mL
• 360 GC Enhancer, 1 x 1.5 mL

Store at -15°C to -25°C.

常見問答集 (常見問題)

My oligonucleotide does not appear to be the right length when I checked by gel electrophoresis. Why is this?

Oligos should be run on a polyacrylamide gel containing 7 M urea and loaded with a 50% formamide solution to avoid compressions and secondary structures. Oligos of the same length and different compositions can electrophorese differently. dC's migrate fastest, followed by dA's, dT's, and then dG's. Oligos containing N's tend to run as a blurry band and generally have a problem with secondary structure.

The primers I am using worked for PCR initially, but over time, have stopped working. What happened?

Primers should be aliquoted for single use before PCR set-up. Heat just the aliquoted primers to 94 degrees for 1 min. Quick chill the primer on ice before adding to the PCR reaction. Some primers may anneal to themselves or curl up on themselves.

I don't see a pellet in my oligo tube order. Should I ask for a replacement?

The drying method dries the primer in a thin layer along the sidewalls of the tube instead of the bottom, therefore a pellet is not always visible and should still be ready to use.

There is a ball-shaped pellet at the bottom of my oligo tube. What is this and can I still use my oligo?

If the oligo was overheated, it will appear as a “ball”-shaped pellet attached to the bottom of the tube. This should not affect the quality of the oligo, and the oligo should be readily soluble in water.

There is a green color in my lyophilized oligo. Can I still use it?

If an oligo appears green in color, this is most likely due to ink falling into the tube. The oligo should still be fully functional. The color can be removed by doing an ethanol precipitation.