Covaris S2/S220 System Tubes & Caps

Catalog number: 4399054

Applied Biosystems™  Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Borosilicate round bottom tubes (13 mm x 65 mm) and caps for use with the Covaris™ S2 System.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Covaris™ S2 System
Format: Tube(s)
Product Line: Applied Biosystems™, Covaris™
Product Size: 125 tube
Type: Cap, Tubes & Vials

Documents

Manuals & protocols