Imidazo[1,2-a]pyridine, 99%
Imidazo[1,2-a]pyridine, 99%
Imidazo[1,2-a]pyridine, 99%
Thermo Scientific Chemicals

Imidazo[1,2-a]pyridine, 99%

CAS: 274-76-0 | C7H6N2 | 118.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 439940050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS274-76-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow to brown
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only