Product Image
Invitrogen™
PrimeFlow™ 96-well plate 10 packets
Polystyrene, v-bottom plates that have been validated for use with the PrimeFlow™ RNA Assay.
Have Questions?
Catalog NumberQuantity
44-17005-4610 Packets
Catalog number 44-17005-46
Price (CLP)
138.178
10 Packets
Quantity:
10 Packets
Price (CLP)
138.178
10 Packets
These PrimeFlow™ 96-well plates are polystyrene, v-bottom plates that have been validated for use with the PrimeFlow™ RNA Assay.
Reported Application
RNA Flow Cytometry
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodColorimetric
Packaging10 Pk.
Plate BlockingNone
Product TypeMicroplate
Quantity10 Packets
ColorClear
MaterialPolystyrene
No. of Wells96
Product LinePrimeFlow
Well ShapeConical Bottom (V-well)
Unit Size10 Packets
Frequently asked questions (FAQs)
Can the FlowRNA Assay be adapted to a 96-well protocol? Do you have a protocol for this?
Yes. We have a validated protocol for Flow RNA assay in 96-wells. Please see Appendix 7 of Cat. No. 88-18005 for the protocol.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
When running the QuantiGene FlowRNA (methanol based) assay, which fluorochromes are recommended for antibody staining?
We recommend the use of fluorochromes that are known to be methanol resistant. Avoid protein-based fluorochromes such as APC, PE, and PerCP and their respective tandems. This question is not appropriate for PrimeFlow RNA Assay.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
For the PrimeFlow RNA Assay, how would you recommend I set compensation?
We recommend using the PrimeFlow Compensation Kit included in the PrimeFlow RNA Assay (Cat. No. 88-18005-204, 88-18005-210) to set compensation for each RNA channel. Refer to Appendix 3 of the PrimeFlow RNA Assay User Manual for detailed instructions for use. The UltraComp eBeads microspheres included in the compensation kit may also be used for any experimental antibodies being used. Antibodies with the same fluorochromes as the RNA probes should not be used to set compensation for RNA probes, as the compensation values will be different.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
I have a 37 degrees C tissue culture incubator in the lab. Can I use this incubator for the hybridization steps in the PrimeFlow RNA Assay protocol?
The hybridization temperature is a critical parameter of this protocol necessary to obtain positive results. The incubator to be used must be validated before use with the ViewRNA Temperature Validation Kit (Cat. No. QV0523), following the protocol in the FlowRNA user manual. Temperatures deviated outside of this range will lead to less-than-optimal hybridization.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
What type of 96-well plate is compatible with the PrimeFlow RNA protocol?
We recommend polystyrene uncoated v-bottom 96-well plates with a lid (Cat. No. 44-17005-46). The u-bottom plate is also acceptable, but flat bottom plates are not recommended.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.