Applied Biosystems™

IKA™ Template Preparation Tubes

Catalog number:  4400401

 Related applications: PCR | SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

These customized tubes and caps are for use with the ULTRA-TURRAX® Tube Drive from IKA® for emulsion PCR for use with the SOLiD system and 5500 series Genetic Analyzers

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer, ULTRA-TURRAX® Tube Drive from IKA®
Format: Tube(s)
Product Line: IKA®
Product Size: 10 tubes
Shipping Condition: Room Temperature
Type: Tubes & Vials

Contents & storage

Each package contains 10 tubes.

Documents

Manuals & protocols