1-Bromo-2-naphthoic acid, 98%
1-Bromo-2-naphthoic acid, 98%
1-Bromo-2-naphthoic acid, 98%
Thermo Scientific Chemicals

1-Bromo-2-naphthoic acid, 98%

CAS: 20717-79-7 | C11H7BrO2 | 251.08 g/mol
Have Questions?
產品號碼QuantityPackaging
4403900101 gGlass bottle
產品號碼 440390010
價格 (TWD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS20717-79-7
規格Specification Sheet規格表
Appearance (Color)White to yellow to light brown
Appearance (Form)Powder
HPLC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only