3500 Genetic Analyzer 8-Capillary Array, 36 cm
Applied Biosystems™

3500 Genetic Analyzer 8-Capillary Array, 36 cm

내부 코팅이 되지 않은 capillary가 array 당 8개씩 세트로 공급되며 built-in frame으로 설치가 용이합니다.36 cm capillary 적용:HID⁄ Forensic Applications.이 제품은자세히 알아보기
Have Questions?
카탈로그 번호수량
44046831 array
카탈로그 번호 4404683
제품 가격(KRW)
2,687,000
온라인 행사
Ends: 30-Jun-2026
2,985,000
할인액 298,000 (10%)
Each
수량:
1 array
제품 가격(KRW)
2,687,000
온라인 행사
Ends: 30-Jun-2026
2,985,000
할인액 298,000 (10%)
Each
내부 코팅이 되지 않은 capillary가 array 당 8개씩 세트로 공급되며 built-in frame으로 설치가 용이합니다.

36 cm capillary 적용:
HID⁄ Forensic Applications.

이 제품은 연구용으로만 사용해야 합니다. 진단 절차에 사용할 수 없습니다.
For Research Use Only. Not for use in diagnostic procedures.
사양
캐필러리 길이36 cm
설명3500 Genetic Analyzer 8-Capillary Array, 36cm Capillary, Internally Uncoated Capillaries, For Use With 3500 Genetic Analyzer, 15 to 30°C Storage Temperature
용도(애플리케이션)Human Identification, Sequencing, Fragment Analysis
용도(장비)3500 Genetic Analyzer
길이(미터법)36 cm (Capillary)
제품 유형Genetic Analyzer 8-Capillary Array
수량1 array
배송 조건Room Temperature
Unit SizeEach
구성 및 보관
Store this array at ambient temperature. For details about storing the capillary array on and off the instrument, please refer to the Applied Biosystems 3500/3500xL Genetic Analyzer User Guide.

자주 묻는 질문(FAQ)

How can I set up auto-analysis for data in GeneMapper with the 3500 Data Collection Software version 3.x?

Beginning with 3500 Data Collection Software version 3, the 3500 Data Collection Software does not support automatic secondary analysis. This is also indicated in the software release notes. To perform secondary data analysis, you will need to manually import the .fsa file into the GeneMapper software. For optimal performance, we recommend that you install the GeneMapper Software on a stand-alone computer that is not part of the 3500 Genetic Analyzer system.

Find additional tips, troubleshooting help, and resources within our Fragment Analysis Software Support Center.

My array is stuck in the pump on my Applied Biosystems 3500/3500xL Genetic Analyzer, even after loosening the knob. What can I do?

If the array is stuck in the pump, even after loosening the knob, squirt around the edge of the array head with distilled water and try to remove it again. If the array will not move, a service call may be required. Do not use excessive force or tools to try and remove it.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.

When I started my run on the Applied Biosystems 3500/3500xL Genetic Analyzer, an error occurred: "Unstable Electrophoresis current detected, check for air bubbles" but I don't see any bubbles. What else can cause this?

Some of the other causes of an Unstable Electrophoresis current detected error message are:

1.Leak on the system
2. Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3. Running Buffer that was:
-On the instrument longer than 2 weeks
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues

Inspect the system for leaks. If you do not see any leaks on the system, perform the wash the pump chamber and channels wizard using the conditioning pouch and place fresh, non-expired polymer and fresh Anode and Cathode buffer containers. If the problem persists, a service call may be required.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.

I have an Applied Biosystems 3500/3500xL Genetic Analyzer. When do my consumables expire?

If we do not specify the expiry date or number of uses, the warranty will last for 12 months from the date of shipment.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.

How often should I change the array on my Applied Biosystems 3500/3500xL Genetic Analyzer?

The Applied Biosystems 3500/3500xl Genetic Analyzer capillary arrays are warrantied for a specific number of runs or until the expiration date printed on the array package, whichever comes first. If you notice that you are starting to get reduced data quality (e.g., shorter reads, trailing peaks in all 4 colors) even with the Sequencing Standard, these may be signs that the array requires changing.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.