3500xL Dx Capillary Array, 36 cm
Applied Biosystems™
3500xL Dx Capillary Array, 36 cm
These internally uncoated 36-cm capillaries are supplied in assemblies of 24 capillaries per array, with a built-in frame for easy installation.
Have Questions?
Catalog NumberQuantity
44046861 Array
Catalog number 4404686
Price (USD)
0.00
Each
Quantity:
1 Array
Price (USD)
0.00
Each
These internally uncoated 36-cm capillaries are supplied in assemblies of 24 capillaries per array with a built-in frame for easy installation. A radio-frequency identification (RFID) tag is built into the frame for tracking critical run information, which is then reported on the software dashboard. The array can be used for 160 injections.
This product is intended for in-vitro diagnostic use and is designed for use with the 3500 Dx and 3500xl Dx genetic analyzers and is not validated for use with other systems or applications.
The 3500 Dx and 3500xl Dx genetic analyzers are to be used by trained operators in the analysis of human DNA for the detection of fluorescently labeled DNA nucleotides by capillary electrophoresis. Refer to the appropriate Applied Biosystems Dx genetic analyzer user guide for further details.
Note: The consumables are for distribution and use in specific European countries only. Not for use in the USA.
  • These internally uncoated 36 cm capillaries are supplied in preassembled sets assemblies of 24 capillaries per array, with a built-in frame for easy installation.
  • Radio frequency identification (RFID) tag is built into the frame for tracking critical run information, which is then reported on the software dashboard. The arrays may be used for 160 injections.
  • This product is intended for in-vitro diagnostic use on the Applied Biosystems 3500 Dx Series Genetic Analyzers and is not validated for use with other systems or applications.
  • The 3500 Dx Series Genetic Analyzer is to be used by trained operators in the analysis of human DNA or RNA, for the detection of genetic changes that may lead to disease presence or susceptibility.
  • Off-label uses such as fragment analysis are for Research Use Only.
  • The 3500 Dx Series Systems and consumables are for distribution and use in specific European countries only. Not for use in the USA.
For In Vitro Diagnostic Use Only
Specifications
Capillary Length36 cm
For Use With (Equipment)3500xL Dx Genetic Analyzer
Quantity1 Array
Shipping ConditionRoom Temperature
Starting MaterialDNA, RNA
Unit SizeEach
Contents & Storage
Ambient, 1 array with 24 capillaries