3500xL Dx Genetic Analyzer 24-Capillary Array, 50 cm
Applied Biosystems™
3500xL Dx Genetic Analyzer 24-Capillary Array, 50 cm
A radio frequency identification tag is built into the frame for tracking critical run information, which is then reported on the software dashboard.
Have Questions?
Catalog NumberQuantity
44046881 Array
Catalog number 4404688
Price (USD)
3,575.00
Each
Quantity:
1 Array
Price (USD)
3,575.00
Each
These internally uncoated 50-cm capillaries are supplied in assemblies of 24 capillaries per array, with a built-in frame for easy installation. Radio frequency identification (RFID) tag is built into the frame for tracking critical run information, which is then reported on the software dashboard. The array can be used for 160 injections in Dx mode.
This product is intended for in vitro diagnostic use on the 3500xL Dx Genetic Analyzer and is not validated for use with other systems or applications. Refer to the 3500 Dx Genetic Analyzer User Guide for further details.
These internally uncoated 50cm capillaries are supplied in preassembled sets assemblies of 24 capillaries per array, with a built-in frame for easy installation.
For In Vitro Diagnostic Use Only
Specifications
Capillary Length50 cm
For Use With (Application)Sequencing
For Use With (Equipment)3500xL Dx Genetic Analyzer, 3500 Genetic Analyzer
Product Type3500xL Dx Genetic Analyzer
Quantity1 Array
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Ambient, 1 array with 24 capillaries
Frequently asked questions (FAQs)
When I started my run on the Applied Biosystems 3500/3500xL Genetic Analyzer, an error occurred: "Unstable Electrophoresis current detected, check for air bubbles" but I don't see any bubbles. What else can cause this?
Some of the other causes of an Unstable Electrophoresis current detected error message are:
1.Leak on the system
2. Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3. Running Buffer that was:
-On the instrument longer than 2 weeks
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues
Inspect the system for leaks. If you do not see any leaks on the system, perform the wash the pump chamber and channels wizard using the conditioning pouch and place fresh, non-expired polymer and fresh Anode and Cathode buffer containers. If the problem persists, a service call may be required.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
Do I need to replace the buffer every 14 days if the Applied Biosystems 3500/3500xL Genetic Analyzer is not in use?
If the instrument is not in use, it is not necessary to replace the buffer every 14 days. However, the buffer level needs to remain at the fill line to prevent the capillary or array from drying out. Top off the buffer volume with water if necessary. Replace the anode and cathode buffer containers prior to starting a new run.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.