내부 코팅이 되지 않은 capillary가 array 당 24개씩 세트로 공급되며 built-in frame으로 설치가 용이합니다.50 cm capillary 적용:Sequencing 및 Fragment Analysis자세히 알아보기
Have Questions?
카탈로그 번호
수량
4404689
1 array
카탈로그 번호 4404689
제품 가격(KRW)
3,593,000
온라인 행사
Ends: 31-Dec-2026
3,992,000
할인액 399,000 (10%)
Each
수량:
1 array
제품 가격(KRW)
3,593,000
온라인 행사
Ends: 31-Dec-2026
3,992,000
할인액 399,000 (10%)
Each
내부 코팅이 되지 않은 capillary가 array 당 24개씩 세트로 공급되며 built-in frame으로 설치가 용이합니다.
50 cm capillary 적용: Sequencing 및 Fragment Analysis 어플리케이션.
이 제품은 연구용으로만 사용해야 합니다. 진단 절차에 사용할 수 없습니다.
For Research Use Only. Not for use in diagnostic procedures.
사양
캐필러리 길이50 cm
설명24-Capillary Array, 50cm Capillary, 125 min. Run Time, Internally Uncoated Capillaries, Fragment Analysis, Protein Quality Analysis, Sequencing Applications, For Use With 3500xL Genetic Analyzer, Room Temperature
용도(애플리케이션)Protein Quality Analysis, Fragment Analysis, Sequencing
용도(장비)3500xL Genetic Analyzer
길이(미터법)50 cm (Capillary)
제품 유형Genetic Analyzer 24-Capillary Array
수량1 array
실행 시간125 min
배송 조건Room Temperature
Software TypeFragment Analysis
Unit SizeEach
구성 및 보관
Store this array at ambient temperature. For details about storing the capillary array on and off the instrument, please refer to the Applied Biosystems 3500/3500xL Genetic Analyzer User Guide.
자주 묻는 질문(FAQ)
My array is stuck in the pump on my Applied Biosystems 3500/3500xL Genetic Analyzer, even after loosening the knob. What can I do?
If the array is stuck in the pump, even after loosening the knob, squirt around the edge of the array head with distilled water and try to remove it again. If the array will not move, a service call may be required. Do not use excessive force or tools to try and remove it.
When I started my run on the Applied Biosystems 3500/3500xL Genetic Analyzer, an error occurred: "Unstable Electrophoresis current detected, check for air bubbles" but I don't see any bubbles. What else can cause this?
Some of the other causes of an Unstable Electrophoresis current detected error message are:
1.Leak on the system
2. Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3. Running Buffer that was:
-On the instrument longer than 2 weeks
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues
Inspect the system for leaks. If you do not see any leaks on the system, perform the wash the pump chamber and channels wizard using the conditioning pouch and place fresh, non-expired polymer and fresh Anode and Cathode buffer containers. If the problem persists, a service call may be required.
How often should I change the array on my Applied Biosystems 3500/3500xL Genetic Analyzer?
The Applied Biosystems 3500/3500xl Genetic Analyzer capillary arrays are warrantied for a specific number of runs or until the expiration date printed on the array package, whichever comes first. If you notice that you are starting to get reduced data quality (e.g., shorter reads, trailing peaks in all 4 colors) even with the Sequencing Standard, these may be signs that the array requires changing.
My Applied Biosystems 3500/3500xL Genetic Analyzer consumables have expired, but how long past expiration can I really use them?
Thermo Fisher Scientific can only provide warranty on performance on non-expired reagents. Using reagents past their expiration date is a risk and may compromise your data integrity.