Applied Biosystems™

Protein Expression Lysate Control Kit (Raji)

Catalog number:  4405448

Related applications:

Real Time PCR (qPCR)

| Stem Cell Analysis

Back to top

Description

The Applied Biosystems® Raji Cell Lysate is a preparation from the Raji human B-cell line that has been characterized for use as an expression control lysate in TaqMan® Protein Expression Assay experiments. The cell lysate was prepared as described in the Protein Expression Sample Preparation Kit Protocol (PN 4405785) using the Protein Expression Sample Preparation Kit (PN 4405443). The Raji Protein Expression Lysate Control Kit provides a non-stem cell line lysate that may be used as a positive control for ICAM1 and CTSB expression. The lysate contains protease and phosphatase inhibitors and has not been subjected to denaturing or reducing conditions.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line: TaqMan™
Product Size: 100 reactions
Shipping Condition: Dry Ice
Species: Human

Contents & storage

Includes Lysate Control (store at<-50°C) and Lysate Dilution Buffer (store at -15 to -25°C).