Product Image
Applied Biosystems™

VetMAX™ AIV Reagents

TaqMan™ AIV-Matrix Reagents enable one-step, real-time, RT-PCR amplification of Avian Influenza Virus matrix gene RNA and Xeno™ RNA Internal Positive Control.
Have Questions?
Catalog NumberQuantity
4405543100 reactions
Catalog number 4405543
Price (MXN)
-
Quantity:
100 reactions

Note: The name of this product has changed. It was previously called TAQMAN AIV REAGENTS EACH.

TaqMan™ AIV-Matrix Reagents enable one-step, real-time, RT-PCR amplification of Avian Influenza Virus matrix gene RNA and Xeno™ RNA Internal Positive Control.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
No. of Reactions100 Reactions
Product LineTaqMan
Quantity100 reactions
Shipping ConditionDry Ice
SpeciesAvian
Target Organism ClassAvian Influenza A, Influenza A
For Use With (Application)Animal Health
Unit SizeEach
Contents & Storage
Store at -20°C. Includes RT-PCR buffer, RT-PCR Enzyme Mix, AIV-Matrix TaqMan™ Primer Probe mix, Detection enhancer, and Nuclease-free water.