This product is being discontinued, please find our new and improved product here: SKU #A53170
VetMAX™ PRRSV NA & EU Controls
Applied Biosystems™

VetMAX™ PRRSV NA & EU Controls

VetMAX™ NA and EU PRRSV Controls are used in multiplex, real-time reverse transcription PCR (RT-PCR) to simultaneously detect and subtype both North American (NA) and European (EU) Porcine Reproductive and Respiratory Syndrome Virus (PRRSV).
Have Questions?
Catalog NumberQuantity
44055481 kit
Catalog number 4405548
Price (USD)
189.00
Each
Quantity:
1 kit
Price (USD)
189.00
Each

Note: The name of this product has changed. It was previously called TAQMAN PRRSV AND XENO RNA CTRL EACH.

VetMAX™ NA and EU PRRSV Controls are used in multiplex, real-time reverse transcription PCR (RT-PCR) to simultaneously detect and subtype both North American (NA) and European (EU) Porcine Reproductive and Respiratory Syndrome Virus (PRRSV). XenoRNA™ RNA Control is an internal positive control added to the lysis solution during RNA isolation to distinguish PCR inhibition, reaction failure, or human error from true negatives. In a single-tube, one-step reaction, PRRSV RNA is reverse transcribed into cDNA.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
FormatTube
Quantity1 kit
Shipping ConditionDry Ice
SpeciesSwine/Porcine
Target Organism ClassPorcine Reproductive and Respiratory Syndrome Virus (PRRSV)
For Use With (Application)Animal Health
Label or DyeQuasar 670
Unit SizeEach
Contents & Storage
Control Template(s)