DNA Dilution Buffer
Applied Biosystems™
DNA Dilution Buffer
DNA Dilution Buffer, a component of the ResDNASEQ Residual DNA Quantitation kits, is offered separately here for those kit users who need additional elution buffer.
Have Questions?
Catalog NumberQuantity
4405587C7 mL
Catalog number 4405587C
Price (USD)
73.00
Each
Quantity:
7 mL
Price (USD)
73.00
Each
DNA Dilution Buffer, a component of the ResDNASEQ Residual DNA Quantitation kits, is offered separately here for those kit users who need additional elution buffer.
As with other products in the Pharma Analytics Sample Preparation product line, DNA Dilution Buffer:
  • Is completely supported by our global manufacturing and distribution centers
  • Has a Certificate of Analysis available
  • Has a shelf-life of up to 12 months
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialDilution Buffer
Recommended StorageStore at 2°C to 8°C.
Physical FormLiquid
Quantity7 mL
Unit SizeEach