Dipyrrolidino(N-succinimidyloxy)carbenium hexafluorophosphate, 97%
Dipyrrolidino(N-succinimidyloxy)carbenium hexafluorophosphate, 97%
Dipyrrolidino(N-succinimidyloxy)carbenium hexafluorophosphate, 97%
Thermo Scientific Chemicals

Dipyrrolidino(N-succinimidyloxy)carbenium hexafluorophosphate, 97%

CAS: 207683-26-9 | C13H20F6N3O3P | 411.29 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4405900505 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 440590050
Price (USD)
191.65
Special offer
Online exclusive
Ends: 28-Dec-2026
225.00
Save 33.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
191.65
Special offer
Online exclusive
Ends: 28-Dec-2026
225.00
Save 33.35
Each
Chemical Identifiers
CAS207683-26-9
IUPAC Name1-{[(2,5-dioxopyrrolidin-1-yl)oxy](pyrrolidin-1-yl)methylidene}-1λ⁵-pyrrolidin-1-ylium; hexafluoro-λ⁵-phosphanuide
Molecular FormulaC13H20F6N3O3P
InChI KeyTVWKVGNCIJUXMY-UHFFFAOYSA-N
SMILESF[P-](F)(F)(F)(F)F.O=C1CCC(=O)N1OC(N1CCCC1)=[N+]1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental Analysis36.45 to 39.48 % (C)
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
Elemental Analysis9.81 to 10.63 % (N)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only