N-alpha-FMOC-N-delta-Trityl-L-glutamine, 95%
N-alpha-FMOC-N-delta-Trityl-L-glutamine, 95%
N-alpha-FMOC-N-delta-Trityl-L-glutamine, 95%
Thermo Scientific Chemicals

N-alpha-FMOC-N-delta-Trityl-L-glutamine, 95%

CAS: 132327-80-1 | C39H34N2O5 | 610.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 440640050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS132327-80-1
IUPAC Name2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-[(triphenylmethyl)carbamoyl]butanoic acid
Molecular FormulaC39H34N2O5
InChI KeyWDGICUODAOGOMO-UHFFFAOYNA-N
SMILESOC(=O)C(CCC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Specific optical rotation-12.5° to -15.5° (20°C, 589 nm) (c=1 in DMF)
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Color)White to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only