VetMAX™ NDV Reagents
Applied Biosystems™

VetMAX™ NDV Reagents

Have Questions?
Catalog NumberQuantity
4406874100 reactions
Catalog number 4406874
Price (MXN)
13,500.34
Each
Quantity:
100 reactions
Price (MXN)
13,500.34
Each
Note: The name of this product has changed. It was previously called TAQMAN NDV REAGENTS EACH.

TaqMan™ NDV Reagents enable one-step, real-time, RT-PCR amplification of Newcastle Disease Virus RNA and Xeno™ RNA Internal Positive Control.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
No. of Reactions100 Reactions
Product LineTaqMan
Quantity100 reactions
Shipping ConditionDry Ice
SpeciesAvian
Target Organism ClassNewcastle Disease Virus (NDV)
For Use With (Application)Animal Health
Unit SizeEach
Contents & Storage
Store at -20°C. Includes RT-PCR buffer, RT-PCR Enzyme Mix, NDV-Matrix TaqMan™ Primer Probe mix, and Nuclease-free water.