Adenosine 5'-monophosphate sodium salt hydrate, 98+%
Adenosine 5'-monophosphate sodium salt hydrate, 98+%
Adenosine 5'-monophosphate sodium salt hydrate, 98+%
Thermo Scientific Chemicals

Adenosine 5'-monophosphate sodium salt hydrate, 98+%

CAS: 149022-20-8 ∣ C10H13N5NaO7P ∣ 369.21 g/mol
数量:
1 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 440710010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS149022-20-8
IUPAC Namesodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate
Molecular FormulaC10H13N5NaO7P
InChI KeyFYWLYWMEGHCZAX-GWKNMROSNA-M
SMILES[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
さらに表示
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=98.0 %
Water=<20.0 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only