PrepSEQ™ Rapid Spin Sample Preparation Kit
Applied Biosystems™
PrepSEQ™ Rapid Spin Sample Preparation Kit
The PrepSEQ™ Rapid Spin Sample Preparation Kit was developed for the rapid preparation of DNA from food and environmental samplesMás información
Have Questions?
Número de catálogoCantidad
44077601 Kit
Número de catálogo 4407760
Precio (MXN)
-
Cantidad:
1 Kit
The PrepSEQ™ Rapid Spin Sample Preparation Kit was developed for the rapid preparation of DNA from food and environmental samples for use in PCR amplification reactions. Using a simple spin protocol, PrepSEQ™ Rapid Spin efficiently inactivates PCR inhibitors. For laboratories involved in food and environmental testing, PCR-based methods are rapidly becoming the standard for accurate, sensitive bacterial detection. This has created the demand for a sample preparation method that is fast, cost-effective and capable of extracting high-quality DNA from a broad range of sample types. While such methods have been developed for dedicated, high-throughput laboratories, there is a growing need for methods suitable to low-to-medium throughput applications. The PrepSEQ™ Rapid Spin Kit meets this need.
• Designed for Food Safety and Environmental Monitoring applications
• Supports low-to-medium throughput work loads (up to 100 samples a day)
• Inactivates PCR inhibitors so you will most likely not have to repeat the template preparation step
• Uses a simple spin protocol
Optimized for PCR-based Food Safety and Environmental Monitoring
For laboratories involved in food safety and environmental monitoring, PCR-based methods are rapidly becoming the standard for accurate, sensitive bacterial detection. Use this sample preparation method for fast, cost-effective extraction of high-quality DNA from a broad range of sample types including chocolate, soft cheeses, and soil.
Designed for Laboratories that Prepare up to 100 Samples a Day
Kits exist for dedicated, high-throughput laboratories, but now you have a kit that is suitable for low-to-medium throughput applications. This kit is designed for laboratories that routinely prepare up to 100 samples a day.
Removes PCR Inhibitors
The PrepSEQ™ Rapid Spin Sample Preparation Kit offers a simple, cost-effective way to prepare bacterial DNA from broth cultures. During the extraction process, the media inside the spin column removes PCR inhibitors from the sample, allowing the bacteria to pass through into the sample collection tube where it can be stored or used for PCR.
For Research Use Only. Not for use in diagnostics procedures.
Especificaciones
Tipo de producto finalBacterial DNA
Para utilizar con (aplicación)PCR en tiempo real cuantitativa (qPCR), PCR
Para utilizar con (equipo)Centrifuge
Compatibilidad de alto rendimientoNo compatible con alto rendimiento (manual)
N.º de reacciones100 reacciones
Línea de productosPrepSEQ™
Tipo de productoRapid Spin Sample Preparation Kit
Cantidad1 Kit
Tipo de muestraSwab, Bacteria (Gram -), Bacterial Culture, Food, Water
Cantidad de material de partida750 μL
ObjetivoADN genómico
FormatoSpin Column
Isolation TechnologyCoarse Filtration, Spin Column
Unit SizeEach
Contenido y almacenamiento
This kit includes 100 columns, 100 1.5 mL tubes (store at 22° to 28°C), and Lysis buffer (store at 2° to 8°C). These reagents are sufficient for 100 samples.