BOC-L-Aspartic acid 1-benzyl ester, 98%, Thermo Scientific Chemicals
BOC-L-Aspartic acid 1-benzyl ester, 98%, Thermo Scientific Chemicals
BOC-L-Aspartic acid 1-benzyl ester, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

BOC-L-Aspartic acid 1-benzyl ester, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
4408200505 g
製品番号(カタログ番号) 440820050
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS30925-18-9
IUPAC Name4-(benzyloxy)-3-{[(tert-butoxy)carbonyl]amino}-4-oxobutanoic acid
Molecular FormulaC16H21NO6
InChI KeyLDRWTKQWSXGSTM-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)NC(CC(O)=O)C(=O)OCC1=CC=CC=C1
さらに表示
Appearance (Color)White to off-white
HPLC>=97.5 %
Water=<1.0 %
Appearance (Form)Powder
Infrared spectrumConforms
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only