3500 Dx Series 96-Well Retainer & Base Set
Applied Biosystems™
3500 Dx Series 96-Well Retainer & Base Set
This is a 96-well retainer & base set (standard) designed for use with the 3500 Dx and 3500xL Dx genetic analyzers.
Have Questions?
Catalog NumberQuantity
44102274 Sets
Catalog number 4410227
Price (EUR)
278,25
Online exclusive
371,00
Save 92,75 (25%)
Each
Quantity:
4 Sets
Price (EUR)
278,25
Online exclusive
371,00
Save 92,75 (25%)
Each
This is a retainer and base set (standard) designed for septa retention in 8-tube strip and 96-well plate formats for use with the 3500 Dx and 3500xL Dx genetic analyzers.
Note: The 3500 Dx and 3500xL Dx genetic analyzers and consumables are for distribution and use in select countries only.
  • This is a 96-well retainer & base set (standard) designed for use with the 3500 Dx⁄3500xL Dx Genetic Analyzer.
  • Septa and plate base⁄retainer kits for 8 tube strip, 96 and 384-well plates used with the 3500 Dx Series Genetic Analyzers.
  • The 3500 Dx Series Systems and consumables are for distribution and use in select countries only.
For In Vitro Diagnostic Use Only
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer
Plate Compatibility96-Well Microtiter Plates, MicroAmp™ Optical 96-Well Reaction Plates
Product TypeBase Set
Quantity4 Sets
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Ambient storage.
Each package contains 4 sets.