Product Image
Applied Biosystems™

Septa for 8-Well Strips, for 3500/SeqStudio™ Flex

These Septa for 8-strips were designed and validated for use with Applied Biosystems 3500 and SeqStudio Flex Series Genetic Analyzers.
Have Questions?
Catalog NumberQuantity
441070124 each
Catalog number 4410701
Price (USD)
709,55
Each
Quantity:
24 each
Price (USD)
709,55
Each

These septa for 8-tube strips were designed and validated for use with Applied Biosystems 3500 and SeqStudio Flex series genetic analyzers.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Equipment)3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers
Format8-well Septa Strip
Product TypeSepta Strip
Quantity24 each
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
This product contains 24 septa.

Frequently asked questions (FAQs)

When I started my run on the Applied Biosystems 3500/3500xL Genetic Analyzer, an error occurred: "Unstable Electrophoresis current detected, check for air bubbles" but I don't see any bubbles. What else can cause this?

Some of the other causes of an Unstable Electrophoresis current detected error message are:

1.Leak on the system
2. Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3. Running Buffer that was:
-On the instrument longer than 2 weeks
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues

Inspect the system for leaks. If you do not see any leaks on the system, perform the wash the pump chamber and channels wizard using the conditioning pouch and place fresh, non-expired polymer and fresh Anode and Cathode buffer containers. If the problem persists, a service call may be required.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.

Do I need to replace the buffer every 14 days if the Applied Biosystems 3500/3500xL Genetic Analyzer is not in use?

If the instrument is not in use, it is not necessary to replace the buffer every 14 days. However, the buffer level needs to remain at the fill line to prevent the capillary or array from drying out. Top off the buffer volume with water if necessary. Replace the anode and cathode buffer containers prior to starting a new run.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.