(R)-alpha-Methyltryptophan hemihydrate, 98%, 98% ee
(R)-alpha-Methyltryptophan hemihydrate, 98%, 98% ee
(R)-alpha-Methyltryptophan hemihydrate, 98%, 98% ee
Thermo Scientific Chemicals

(R)-alpha-Methyltryptophan hemihydrate, 98%, 98% ee

CAS: 56452-52-9 | C24H30N4O5 | 454.53 g/mol
Have Questions?
Catalog NumberQuantity
441191000100 mg
Catalog number 441191000
Price (USD)
-
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS56452-52-9
IUPAC Namebis(2-amino-3-(1H-indol-3-yl)-2-methylpropanoic acid) hydrate
Molecular FormulaC24H30N4O5
InChI KeyBZCFHAYHUKOGKE-IUWNWRFPSA-N
SMILESO.CC(N)(CC1=CNC2=CC=CC=C12)C(O)=O.CC(N)(CC1=CNC2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Enantiomeric excess>=98.0 %
Specific optical rotation+14° to +18° (20°C, 589 nm) (c=0.9 in water)
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only