The MagMAX beads will cause a loss of ~2-3 µL of solution when they are pulled from the solution, but this is a very small amount. Larger volume loss, such as 20µL, can be due to several factors:
• Low lab humidity combined with a heated elution can cause this amount of liquid to evaporate. In this case, we recommend increasing the starting elution volume. Since the nucleic acid is not evaporating this should not affect your overall yield.
• Issues with alignment of the instrument components could also result in loss of volume. If the head goes down too deep during mixing, it could pull droplets up along the side. This would reduce elution volume and you would also be more likely to see a reduction in yield. A similar issue would result if the turntable alignment was off on either side.
Find additional tips, troubleshooting help, and resources within our
Automated Nucleic Acid Purification Support Center.