I'm using your MagMAX DNA Multi-Sample Isolation Kit for isolation of genomic DNA, but I've run out of DNA binding beads. Can I order them as a standalone product?
Yes, please order Cat. No. 4489112.
Search
View additional product information for MagMAX™-96 DNA Multi-Sample Kit - FAQs (4413022, 4413021)
7 product FAQs found