MagMAX™-96 DNA Multi-Sample Kit
Invitrogen™

MagMAX™-96 DNA Multi-Sample Kit

MagMAX™ DNA Multi-Sample 96-Well Kits are designed for rapid purification of high-quality genomic DNA from a variety of sample types including blood and blood cards, tissue, mouse tails, buccal swabs, buffy coats, and cultured cells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
4413022480 Preps
441302196 Preps
Catalog number 4413022
Price (USD)
2,505.00
Each
No. of Reactions:
480 Preps
Price (USD)
2,505.00
Each

MagMAX™ DNA Multi-Sample 96-Well Kits are designed for rapid purification of high-quality genomic DNA from a variety of sample types including blood and blood cards, tissue, mouse tails, buccal swabs, buffy coats, and cultured cells. These purification kits use MagMAX™ magnetic bead-based nucleic acid isolation technology to produce high yields of purified DNA, free from inhibitors that may affect downstream reactions such as genotyping assays, CE and next-generation sequencing, and high resolution melting analysis. The 96-prep and 5 x 96-prep kits are designed for higher throughput and automated format processing.

The demands for DNA yield and purity have become more stringent with the development of new analytical methods and technologies, such as high-throughput genotyping, and next-generation DNA sequencing platforms. MagMAX™ DNA Multi-Sample Kits meet the challenge by delivering highly pure DNA that is free of nucleases, proteins, and other inhibitors of downstream enzymatic reactions.

As shown in Table 1, DNA with high yield and purity can be obtained from a variety of materials using the MagMAX™ DNA Multi-Sample Kits. This is in contrast to filter membrane–based methods that often require different kit configurations for various input materials. For applications requiring maximal yields, for small batch sizes, or for DNA archival purposes MagMAX™ DNA Multi-Sample Kit (tube format, manual processing) may be desirable because larger sample inputs are accommodated. DNA yields suitable for most routine applications may be obtained using the MagMAX™-96 DNA Multi-Sample Kits (96-well plate format, compatible with automated handling) (Table 1).

The MagMAX™ DNA Multi-Sample Kit also delivers high yields of DNA from lipid or glycogen rich samples, such as brain and liver tissues, respectively. Figure 1 shows more than twice as much DNA was purified from tissues with the MagMAX™ DNA Multi-Sample Kit than with the leading competitor’s filter membrane–based method.

Most enzymatic molecular applications are prone to inhibition by analytes carried over from the sample matrix (e.g., heme, heparin, salts, protein) or the purification method itself (phenol, salts, alcohol). Figure 2 illustrates the effect of DNA purity on sensitive molecular applications, such as quantitative PCR (qPCR). DNA was extracted from blood using the MagMAX™-96 DNA Multi-Sample Kit, or a competitor’s filter membrane–based kit and subjected to real-time PCR analysis. The cycle threshold CT values obtained with the DNA purified using the MagMAX™ DNA Multi-Sample Kit were lower than those obtained with the filter membrane kit–purified DNA, indicating the MagMAX™ kit yields higher-purity DNA.

MagMAX™ DNA Multi-Sample Kits provide purified DNA that is suitable for a variety of Applied Biosystems application workflows (Figure 3), including SNP genotyping and DNA sequencing analysis (see Figure 4). To see the full performance description of DNA purified using the MagMAX™ DNA Multi-Sample Kits in TaqMan™ SNP genotyping assays and with BigDye™ chemistry in DNA sequence analysis, view the MagMAX™ DNA Multi-Sample Kit Product Bulletin.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Final Product TypeGenomic DNA
For Use With (Application)Genotyping, Real-Time Quantitative PCR (qPCR), PCR, Sequencing
High-throughput CompatibilityHigh-throughput Compatible, Not High-throughput Compatible (Manual), Automated Protocols
No. of Reactions480 Preps
Product LineMagMAX
Product TypeDNA Multi-Sample Kit
Quantity5 x 96 Preps
Sample TypeBlood, Buccal Samples, Cells, Tissue
TargetGenomic DNA
FormatKit
Isolation TechnologyMagnetic Bead
Unit SizeEach
Contents & Storage
Contains:
• DNA Binding Beads, 10 mg/mL, 1 x 8.5 mL. Store at 2° to 8°C
• DNA Elution Buffer 1, 1 x 72 mL. Store at RT
• DNA Elution Buffer 2, 1 x 72 mL. Store at RT
• Elution Plate, 96-well, 3 plates. Store at RT
• Wash Solution 1 Concentrate, 1 x 75 mL (for PN 4413021). Store at RT
• Wash Solution 2 Concentrate, 1 x 250 mL. Store at RT
• Multi-Sample DNA Lysis Buffer, 1 x 250 mL. Store at RT
• MicroAmp™ Clear Adhesive Film, 5 x 4 films. Store at RT
• Nuclease-Free Water, 2 x 50 mL. Store at RT
• PK Buffer 1 x 50 mL, Store at RT
• Processing Plate, 96-well, 5 plates. Store at RT
• Proteinase K, 1 x 4.3 mL. Store at -15° to -25°C
• Ribonuclease A (RNase A), 1 x 2.7 mL. Store at -15° to -25°C

Frequently asked questions (FAQs)

I'm using your MagMAX DNA Multi-Sample Isolation Kit for isolation of genomic DNA, but I've run out of DNA binding beads. Can I order them as a standalone product?

Yes, please order Cat. No. 4489112.

Can I edit the manufactured KingFisher Flex protocols?

No, manufactured protocols are locked for editing, but they can be renamed using the ‘save as' function and edited as a new version.

Can I use magnetic beads from other vendors with the KingFisher Flex instrument?

For nucleic acid extraction, we recommend using our MagMAX kits and MagJET kits. Other magnetic beads should work well as long as they are 0.5 to 10 µm in size. However, you will have to write your own KingFisher protocols using the Thermo Scientific BindIt software and validate/optimize the protocol on your own.

Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.

I am working with liquid samples with high volume and want to extract nucleic acid from them using the KingFisher Flex instrument. Can I use more volume than the protocol advises?

Yes, this can be done by sample splitting. Set the protocol so that the magnet will collect samples from multiple wells before continuing to the next step. Be sure not to overload.

Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.

Can I scale up my reaction volume with the KingFisher Flex instrument?

Yes, reaction volumes can be scaled up, and all other volumes (i.e. wash volumes) will have to be increased by the same fold difference. It is generally not recommended to scale up reaction volumes more than 2-fold unless the plate's volume capacity will allow it.