resDNASEQ™ Quantitative CHO DNA Kits
Applied Biosystems™

resDNASEQ™ Quantitative CHO DNA Kits

This quantitative PCR (qPCR)-based kit, part of the resDNASEQ™ Quantitative CHO DNA System, allows users to detect sub-picogram amounts of residual DNA from CHO cell line in under 5 hours.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
4413713Assay Kit and Sample Preparation
4402085Assay Kit only
Catalog number 4413713
Price (MXN)
101,468.80
Each
Includes:
Assay Kit and Sample Preparation
Price (MXN)
101,468.80
Each
This quantitative PCR (qPCR)-based kit, part of the resDNASEQ™ Quantitative CHO DNA System, enables you to detect sub-picogram amounts of residual DNA from the Chinese hamster ovary (CHO) cell line in under 5 hours. The system overcomes the limitations of traditional methods by combining high-recovery PrepSEQ™ sample preparation and TaqMan™-based quantitation of residual DNA.
Real-time quantitative PCR kit for highly sensitive residual CHO DNA quantitation
• Easy to use, results in under 5 hours
• Specific to CHO DNA, no cross-reactivity with unrelated DNA
• Part of an integrated system that includes sample preparation, TaqMan™ assay and master mix, standard DNA, instrument, and software
• Protocol available for automated sample preparation
• Flexible sample throughput allows you to perform rigorous DNA clearance studies

This kit is available with or without sample preparation reagents in the form of a PrepSEQ Residual DNA Sample Preparation Kit, which is optimized for highly efficient residual DNA recovery from complex mixtures of proteins, buffers, and salts.

Real-Time PCR for Highly Sensitive Quantitation
The resDNASEQ™ Quantitative CHO DNA System provides highly sensitive detection of CHO DNA, allowing you to use small sample volumes to generate accurate results. The broad linear range of TaqMan™ technology allows you to test samples with variable levels of CHO DNA in the same assay, such as in-process samples with higher amounts of DNA or bulk drug substance with very low amounts. Figure 1 demonstrates the range and sensitivity of the assay. Linearity is demonstrated by analysis of CHO cell standard DNA ranging from 0.3 ng to 3 fg.

Easy to Use, Results in Under 5 Hours
First, you extract DNA from your in-process purification and drug substance samples. Then you perform qPCR to compare DNA amounts in your test samples to a standard curve generated with known amounts of purified CHO cell standard DNA. The easy workflow of the resDNASEQ™ Quantitative CHO DNA System consists of sample preparation, assay setup, and instrument run and data analysis—all of which you can perform in under 5 hours.

Specific to CHO DNA, No Cross-Reactivity With Unrelated DNA
The target of the assay is highly specific, so that it only detects a hamster-specific region of a multicopy genetic element. We selected this region using extensive bioinformatic analysis of multiple related and unrelated species. Testing of assay performance confirmed that the assay is specific to hamster DNA and is unaffected by the presence of as much as 100 ng of unrelated DNA in a test sample.

Consistent Performance Even With Fragmented DNA
For accurate quantitation of residual CHO DNA, assay results must be unaffected by the size of the DNA molecules present in the test sample. To test the effect of DNA fragment size on assay performance, we fragmented high molecular weight CHO genomic DNA into low molecular weight DNA by sonication. Figure 2 demonstrates that the threshold cycle (Ct) values for the reactions with the sonicated low molecular weight DNA were comparable to those of the undigested high molecular weight DNA. These results demonstrate that consistent performance was obtained with the kit irrespective of DNA molecular weight.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatKit
Host CellCHO
IncludesAssay Kit and Sample Preparation
No. of Reactions100 Reactions
Product LineresDNASEQ
Quantity100 reactions
Detection MethodPrimer-probe
Label or DyeFAM
PCR MethodqPCR
Unit SizeEach
Contents & Storage
resDNASEQ CHO DNA Control
• CHO DNA Control, 30 ng/μL, 40 μL, store at –15 to –25°C
• DNA Dilution Buffer (DDB), 7 mL, store at –15 to –25°C before first use and 2−8°C after first use

resDNASEQ CHO DNA Real-Time PCR Reagents
• Environmental Master Mix (2X), 2 x 0.75 mL, store at –15 to –25°C before first use and 2−8°C after first use, protect from light
• CHO DNA Assay Mix (10X), 300 μL, store at –15 to –25°C, protect from light
• Negative Control (water), 1.0 mL, store at –15 to –25°C before first use and 2−8°C after first use

PrepSEQ Nucleic Acid Extraction Kit
• Lysis Buffer (2 bottles, 50 mL/bottle)
• Binding Solution/Isopropanol (1 empty bottle)
• Wash Buffer Concentrate (2 bottles, 26 mL/bottle)
• Elution Buffer (1 bottle, 25 mL)
• Proteinase K Buffer (1 bottle, 50 mL)
• Magnetic Particles (2 tubes, 1.5 mL/tube)
• Proteinase K (20 mg/mL) (1 tube, 1.25 mL), store at or below –20°C
• PK Buffer II (1 bottle, 11 mL)
Store all at room temperature unless otherwise indicated.

PrepSEQ Residual DNA Sample Preparation Kit
• Proteinase K (20 mg/mL), 1 tube, 1.25 mL
• Yeast tRNA (10 mg/mL), 1 tube, 0.5 mL
• Glycogen (5 mg/mL), 2 tubes, 1.0 mL/tube
Store all at or below –20°C.