Tetrakis(triphenylphosphine)palladium(0), 99.9%, (trace metal basis)
Tetrakis(triphenylphosphine)palladium(0), 99.9%, (trace metal basis)
Tetrakis(triphenylphosphine)palladium(0), 99.9%, (trace metal basis)
Thermo Scientific Chemicals

Tetrakis(triphenylphosphine)palladium(0), 99.9%, (trace metal basis)

CAS: 14221-01-3 | C72H60P4Pd | 1155.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 441400010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS14221-01-3
IUPAC Nametetrakis(triphenylphosphane) palladium
Molecular FormulaC72H60P4Pd
InChI KeyNFHFRUOZVGFOOS-UHFFFAOYSA-N
SMILES[Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Yellow to yellow-green to orange
Palladium (Pd)8.8 to 9.6 %
Total trace metal impurities=<0.1 %
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only