Product Image
Applied Biosystems™

TaqMan™ Array Human cAMP Pathway

These 96-well plates are pre-configured with the most appropriate TaqMan® Gene Expression Assays for a specific biological process, pathway, or자세히 알아보기
Have Questions?
카탈로그 번호수량
44141141 standard plate
카탈로그 번호 4414114
제품 가격(KRW)
369,000
Each
수량:
1 standard plate
제품 가격(KRW)
369,000
Each
These 96-well plates are pre-configured with the most appropriate TaqMan® Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The TaqMan® Array Human cAMP Pathway 96-well Plate contains 92 assays to genes associated with cAMP pathways and 4 assays to candidate endogenous control genes.
For Research Use Only. Not for use in diagnostic procedures.
사양
형식96-well plate
내부 프로브 수정MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
반응 수1 x 20 μL Reactions
제품라인TaqMan
수량1 standard plate
연구 카테고리Cell Cycle & Proliferation, Signal Transduction, Biomarker-Related Pathway(s), Immunology, Oncology
종Human
형태Dried Down
라벨 또는 염료FAM
반응 속도Standard
Unit SizeEach
구성 및 보관
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 20 μL reaction. The plates can be stored at room temperature or at 4°C.

자주 묻는 질문(FAQ)

What is the minimum amount of TaqMan array plates I need to purchase to submit an order?

The minimum amount of TaqMan Array plates needed to submit an order is based on the plate type:
• Inventoried TaqMan array plates: 1 plate
• TaqMan Flexible Content panel array plates: 6 plates
• Custom TaqMan array plates: 6 plates
• Specialty TaqMan array plates: 10-20 plates (depends on assay format and configuration). Please contact our Specialty Plates team at Specialty_Plates@thermofisher.com for details.

Can our TaqMan Array Plates be run on non-Applied Biosystem real-time PCR instruments?

We recommend using our Plastics Selection Tool to verify which plates are compatible with your instrument model, https://www.thermofisher.com/us/en/home/life-science/pcr/pcr-plastics/plastics-selection-guide.html.

Based on our testing, the 96-well TaqMan Array Plates can be used on the following non-Applied Biosystem real-time PCR instruments:
• Standard (0.2mL) 96-well Plates: Bio-Rad IQ5
• Fast (0.1mL) Plates: Bio-Rad CFX Connect and Bio-Rad CFX96 Touch

How do I obtain TaqMan documentation files for my TaqMan array plates?

Beginning in April 2016, TaqMan documentation files for TaqMan Array Plates (assay information files (AIF), experimental setup files, and plate layout files) can be downloaded from the Thermo Fisher Scientific website at https://www.thermofisher.com/taqmanfiles. To obtain your TaqMan documentation files:
• Select the appropriate product type.
• Enter the information being requested for the product type.
• Fixed TaqMan array plates: Enter the Part number and Batch number
• TaqMan Flexible Content panel array plates/Custom TaqMan array plates: Enter the Sales Order number and Batch number
• Click Submit.