AmpFLSTR® NGM™ PCR Amplification Kit

Catalog number: 4415021

Applied Biosystems™  Related applications: Forensics

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The NGM™ Kit is a highly robust Short Tandem Repeat (STR) multiplex kit which utilizes a single protocol to amplify the 10 SGM Plus™ loci (D2S1338, D3S1358, D8S1179, D16S539, D18S51, D19S433, D21S11, FGA, TH01, vWA, Amelogenin) together with the additional loci approved by the European Union Council for the expansion of the European Standard Set (D10S1248, D22S1045, D2S441, D1S1656 & D12S391). The kit combines the latest advances in buffer optimization, primer synthesis and thermal cycling optimization to deliver high sensitivity and improved STR performance for forensic casework and database samples in one easy workflow.

Key Features of the NGM™ Kit:

• Simultaneous amplification of the SGM Plus® kit loci plus 5 additional ENFSI/EDNAP recommended loci to provide powerful discrimination and facilitate cross-border data sharing
• Unprecedented performance in the presence of significant inhibitors combined with enhanced profile and locus balance
• Increased ability to recover information from degraded samples through a concentration of highly discriminatory loci in the low molecular weight region
• Greater sensitivity for improved amplification of low-level samples and better detection of the minor contributor in mixed samples
• Exceptional profile quality and clean baseline resulting from improved primer synthesis and purification processes
• 100% concordance with previously typed samples through maintenance of all STR primer sequences for loci common to the SGM Plus®, Identifiler® and SEfiler Plus™ Kits
• Proprietary and proven mobility modifier technology enables optimal locus spacing
• One kit, one protocol delivers high quality results from both database and casework samples
• Streamlined workflow with reduced amplification time, enzyme pre-formulated within the master mix and a larger package size option (1000 reactions) for high-throughput laboratories
• Optimized as part of an integrated reagent, instrumentation and software system for maximum performance and efficiency
• Outstanding discrimination and robustness offers utility for specialized applications including complex paternity/kinship, disaster victim identification and missing persons cases

Note: For Forensic or Paternity Use Only.

Important Notice

The purchase of this product conveys to the buyer the non-transferable license to use the purchased amount of the product only for forensic or paternity testing conducted by the buyer. This license does not include, as ruled by a U.S. Court, a license to use this product in chimerism determination or analysis, molar specimens classification, cell line authentication, determination of fetal sex, cancer analysis, genetic research, non-casework-related forensic applications (such as general research in forensics or teaching and training persons not employed in a forensic laboratory), identifying or determining maternal cell contamination, or sample tracking. Further rights can be obtained by contacting Promega Corp., 2800 Woods Hollow Rd., Madison, WI 53711 USA.
For Forensic or Paternity Use Only.

Specifications

Amplicon Size: <380bp
For Use With (Equipment): 310 Genetic Analyzer, 3100-Avant™ Genetic Analyzer, 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, GeneAmp® 9600, GeneAmp™ 9700
Label or Dye: FAM™, LIZ™, NED™, PET™, VIC®
Matrix and Size Standards: GeneScan™ 500 LIZ® Size Standard, Matrix Standard Set DS-33
Polymerase: AmpliTaq Gold® DNA Polymerase
Product Line: AmpFLSTR®, NGM™
Product Size: 1,000 reactions
Reaction Format: SuperMix or Master Mix
Sample Type (Specific): DNA (Genomic), Forensic Sample
Sample Volume: 25 µL⁄reaction
Technique: STR (Short Tandem Repeat) Analysis
Shipping Condition: Dry Ice

Contents & storage

• AmpFLSTR® NGM™ Master Mix
• AmpFLSTR® NGM™ Primer Set
• AmpFLSTR® NGM™ Allelic Ladder
Store at ?15 to ?25 °C upon receipt, 2 to 8 °C after initial use

• AmpFLSTR® Control DNA 007
Store at 2 to 8 °C

Documents

Manuals & protocols